Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 16 - 17 | 2039 | 86 | |
AK287031.1 | - | 25 - 26 | 3863 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |