Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249446.1 | NP_001236375.1 | 4 - 5 | 1106 | 96 | |
GenBank mRNA | AJ875407.1 | - | 4 - 5 | 1106 | 96 | |
AK245571.1 | - | 4 - 5 | 1123 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |