Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019782.hg.1 |
ENST00000308064.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019782.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019782.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019781.hg.1 |
ENST00000308064.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019781.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019781.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100180627.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180627.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180627.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180627.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180627.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100180628.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180628.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180628.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180628.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180628.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |