Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000345034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000345034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_006511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_006511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003174.hg.1 |
TC01000193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003174.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003176.hg.1 |
TC01000193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003176.hg.1 |
ENST00000480945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003176.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003175.hg.1 |
TC01000193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003175.hg.1 |
ENST00000480945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003175.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003177.hg.1 |
TC01000193.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003177.hg.1 |
ENST00000480945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003177.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148760.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000320153.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000345034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000345034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000483899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
ENST00000548451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_006511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_006511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006991.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148757.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148759.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148756.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148756.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148756.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148756.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148756.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000480945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000486680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
ENST00000546927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148758.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148755.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148755.hg.1 |
NM_032341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148755.hg.1 |
TC0100006991.hg.1 |
Clariom D TC |