Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000243457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000243457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000243457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000243457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000535240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000535240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
ENST00000535240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
NM_000891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
NM_000891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
NM_000891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008769.hg.1 |
NM_000891.2 |
Refseq |