Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000737.3 | NP_000728.1 | 1 - 2 | 415 | 55 | |
GenBank mRNA | AK291552.1 | - | 1 - 2 | 373 | 55 | |
BC022796.1 | - | 1 - 2 | 353 | 4811294 | 55 | |
BC106059.1 | - | 1 - 2 | 361 | 4992804 | 55 | |
J00117.1 | - | 1 - 2 | 36 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Stem Cell Pluripotency 1700 Inventoried Human_Stem_Cell Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31622611 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 558526 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 640180 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 21533 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 336891 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 128328 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P395814 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P395814 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 40188 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35981 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15171 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P395814 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00328434 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P395814 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31622611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31622611 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31622611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31622611 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P395814 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P41216119 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03459141 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P41216119 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P395814 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17151854 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17151797 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985390 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985395 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985384 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17151785 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17151778 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10985392 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8038302 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038296 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038295 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038289 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038282 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038283 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030190 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038281 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038290 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038294 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030195 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038284 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038301 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038300 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030185 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030193 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030184 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030196 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030192 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030186 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038297 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030191 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8030194 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8038293 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8038299 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8030181 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8038292 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8038288 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8038280 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8030187 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR19023267.hg.1 | TC19002714.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023267.hg.1 | ENST00000357383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023267.hg.1 | ENST00000448402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023267.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023268.hg.1 | TC19002714.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023268.hg.1 | ENST00000357383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023268.hg.1 | ENST00000448402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023268.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206982.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206982.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206982.hg.1 | TC1900011993.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206982.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206982.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | ENST00000301408.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008516.hg.1 | NM_033043.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011993.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206983.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206983.hg.1 | NM_000737.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206983.hg.1 | TC1900011993.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206983.hg.1 | ENST00000357383.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206983.hg.1 | NM_000737.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 205387_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 205387_s_at | - | - |
Human Genome U133A 2.0 Array | 205387_s_at | - | - |
Human Genome U133A 2.0 Array | 205387_PM_s_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw553505 | - | - |
Human Genome U95 Set | 1152_PM_i_at | - | - |
Human Genome U95 Set | 1122_f_at | - | - |
Human Genome U95 Set | 1152_i_at | - | - |
Human Genome U95 Set | 1153_PM_f_at | - | - |
Human Genome U95 Set | 1153_f_at | - | - |
Human Genome U95 Set | 1122_PM_f_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205387_s_at6 | - | - |
Human Genome U133 Set | 205387_s_at | - | - |
Human Genome U133 Set | 205387_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012881 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011674 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011678 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011678 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011674 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012881 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 143219 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 151497 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2153029 | BC103970.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2153029 | BC103971.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2153029 | BC006290.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1683469 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15451751-S | CR593396.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15451751-S | CR613376.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15451751-S | NM_000737.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15451751-S | AK291552.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_18771-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001050239 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001050239 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_18771 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18771 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2153029 | BC051378.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2153029 | BC103970.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2153029 | BC103971.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2153029 | NM_033043.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2153029 | BC006290.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2153029 | BC051378.1 | human_transcriptome |