Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014246.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014246.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014246.hg.1 |
TC16000988.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175929.hg.1 |
TC1600009864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000356897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
ENST00000568075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009864.hg.1 |
NM_145659.3 |
Refseq |