Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318339.1 | NP_001305268.1 | 1 - 2 | 412 | 77 | |
GenBank mRNA | AB023231.2 | - | 1 - 2 | 405 | 77 | |
DA238225.1 | - | 1 - 2 | 394 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |