Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016595.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016595.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016595.hg.1 |
TC12001274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016597.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016597.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016597.hg.1 |
TC12001274.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010018.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175656.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175656.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175656.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175656.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175656.hg.1 |
TC1200010018.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200175654.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175654.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175654.hg.1 |
ENST00000316048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175654.hg.1 |
NM_001013698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175654.hg.1 |
TC1200010018.hg.1 |
Clariom D TC |