Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194291.2 | NP_919267.2 | 4 - 5 | 1013 | 99 | |
XM_011516847.2 | XP_011515149.1 | 4 - 5 | 1007 | 99 | ||
GenBank mRNA | BC017881.1 | - | 2 - 3 | 117 | 99 | |
BC032396.1 | - | 4 - 5 | 314 | 5197382 | 99 | |
BC041379.1 | - | 4 - 5 | 1010 | 5275048 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |