Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017529.hg.1 |
TC04001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017529.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017529.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017531.hg.1 |
TC04001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017531.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017531.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017530.hg.1 |
TC04001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017530.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017530.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011012.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174858.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174858.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174858.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174858.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174858.hg.1 |
TC0400011012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174860.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174860.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174860.hg.1 |
ENST00000296029.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174860.hg.1 |
NM_002619.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174860.hg.1 |
TC0400011012.hg.1 |
Clariom D TC |