Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010591.hg.1 |
ENST00000250930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010591.hg.1 |
ENST00000382942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010591.hg.1 |
ENST00000431232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010591.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010591.hg.1 |
TC16000727.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167504.hg.1 |
TC1600008943.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000250930.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000424078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000427313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000431232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000448854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000467452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000475348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
ENST00000476735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008943.hg.1 |
NM_021259.2 |
Refseq |