Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM47B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152631.2 | NP_689844.2 | 1 | 1662 | 63 | |
GenBank mRNA | AK093101.1 | - | 1 | 1644 | 63 | |
AL043093.3 | - | 1 | 63 | |||
BC035026.2 | - | 1 | 1662 | 4828379 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693004 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11692997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693005 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397893 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17332206 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17332200 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17332202 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11692998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11693013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32397855 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11692996 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 284077 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P105381 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P105381 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 28830 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20036 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03687198 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P105381 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P105381 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17332165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693004 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397846 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693008 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17332165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693004 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397846 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693008 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11692997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11693006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32397848 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P105381 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11692998 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693002 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693004 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11692996 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693000 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693007 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693009 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693003 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693001 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11693005 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P105381 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P105386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693002 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11692998 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693015 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693004 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693014 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693011 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693007 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693001 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693013 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693000 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11693006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_152631_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8166666 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8166665 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 240426_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11731796_at | - | - |
Affymetrix Human Genome U219 Array | 11731796_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147158.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147158.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147158.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147158.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147158.hg.1 | TC0X00006924.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | ENST00000329357.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006924.hg.1 | NM_152631.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 240426_at | - | - |
Human Genome U133 Plus 2.0 Array | 240426_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_152631_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240426_at9 | - | - |
Human Genome U95 Set | 69451_PM_at | - | - |
Human Genome U95 Set | 69451_at | - | - |
Human Genome U133 Set | 240426_PM_at | - | - |
Human Genome U133 Set | 240426_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040818 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040818 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 116209 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2168903 | BC035026.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2168903 | AK093101.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2168903 | AL043093.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778940 | BC035026.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778940 | AK093101.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_22749288-S | BC035026.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_22749288-S | NM_152631.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_22749288-S | AK093101.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_22749288-S | AL043093.3 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_25468-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0006840609 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0006840609 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_25468 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_25468 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2168903 | AL043093.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778940 | BC035026.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778940 | NM_152631.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778940 | AK093101.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778940 | AL043093.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2168903 | BC035026.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2168903 | NM_152631.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2168903 | AK093101.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2168903 | AL043093.3 | human_transcriptome |