Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007911.hg.1 |
ENST00000291041.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007911.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007911.hg.1 |
TC16000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007912.hg.1 |
ENST00000291041.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007912.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007912.hg.1 |
TC16000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007910.hg.1 |
ENST00000291041.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007910.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007910.hg.1 |
TC16000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161119.hg.1 |
TC1600008178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161122.hg.1 |
TC1600008178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161118.hg.1 |
TC1600008178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
ENST00000575198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008178.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
ENST00000291041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
ENST00000570631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
NM_006742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161123.hg.1 |
TC1600008178.hg.1 |
Clariom D TC |