Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004747.3 | NP_004738.3 | 30 - 31 | 5613 | 94 | |
XM_005270276.4 | XP_005270333.1 | 30 - 31 | 5567 | 94 | ||
XM_006718056.3 | XP_006718119.1 | 29 - 30 | 4556 | 94 | ||
XM_011540341.2 | XP_011538643.1 | 30 - 31 | 5737 | 94 | ||
XM_011540342.1 | XP_011538644.1 | 30 - 31 | 5315 | 94 | ||
XM_011540344.2 | XP_011538646.1 | 29 - 30 | 5222 | 94 | ||
XM_011540345.1 | XP_011538647.1 | 29 - 30 | 5347 | 94 | ||
XM_011540347.2 | XP_011538649.1 | 26 - 27 | 4793 | 94 | ||
XM_017016913.1 | XP_016872402.1 | 30 - 31 | 5276 | 94 | ||
XM_017016914.1 | XP_016872403.1 | 30 - 31 | 5606 | 94 | ||
XM_017016915.1 | XP_016872404.1 | 26 - 27 | 5477 | 94 | ||
GenBank mRNA | AB091676.1 | - | 28 - 29 | 5078 | 94 | |
AK314771.1 | - | 14 - 15 | 1922 | 94 | ||
BC073996.1 | - | 16 - 17 | 2323 | 94 | ||
BC117695.2 | - | 20 - 21 | 5371 | 94 | ||
BX649131.1 | - | 25 - 26 | 3721 | 94 | ||
U61843.1 | - | 15 - 16 | 2171 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |