Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000332439.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000570446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000571846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000573940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000574357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
ENST00000624171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006774.hg.1 |
NM_144607.5 |
Refseq |