Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136263.1 | NP_001129735.1 | 1 - 2 | 169 | 119 | |
XM_011527694.1 | XP_011525996.1 | 1 - 2 | 127 | 119 | ||
GenBank mRNA | AB075837.1 | - | 1 - 2 | 169 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |