Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK075140.1 | - | 3 - 4 | 231 | 63 | |
AY358081.1 | - | 3 - 4 | 220 | 63 | ||
AY358283.1 | - | 3 - 4 | 257 | 63 | ||
BC020731.1 | - | 3 - 4 | 231 | 4692272 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |