Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005717.hg.1 |
ENST00000282344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005717.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005717.hg.1 |
TC13000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005716.hg.1 |
ENST00000282344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005716.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005716.hg.1 |
TC13000506.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300156801.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156801.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156801.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156801.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156801.hg.1 |
TC1300008424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156802.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156802.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156802.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156802.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156802.hg.1 |
TC1300008424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000282344.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000612674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
ENST00000620323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008424.hg.1 |
NM_182488.3 |
Refseq |