Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000551.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000551.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000551.hg.1 |
TC20000032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000552.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000552.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000552.hg.1 |
TC20000032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000550.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000550.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000550.hg.1 |
TC20000032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000553.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000553.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000553.hg.1 |
TC20000032.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000144914.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144914.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144914.hg.1 |
TC2000006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144914.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144914.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144910.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144910.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144910.hg.1 |
TC2000006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144910.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144910.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144913.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144913.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144913.hg.1 |
TC2000006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144913.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144913.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006534.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144912.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144912.hg.1 |
NM_030811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144912.hg.1 |
TC2000006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144912.hg.1 |
ENST00000380325.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144912.hg.1 |
NM_030811.3 |
Refseq |