Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024791.hg.1 |
ENST00000392876.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024791.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024791.hg.1 |
TC12001901.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204253.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204253.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204253.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204253.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204253.hg.1 |
TC1200012835.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000392876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000415849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000465502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
ENST00000474268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012835.hg.1 |
NM_001031701.2 |
Refseq |