Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023218.hg.1 |
TC05001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023218.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023218.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023219.hg.1 |
TC05001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023219.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023219.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023216.hg.1 |
TC05001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023216.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05023216.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181851.hg.1 |
TC0500011875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181849.hg.1 |
TC0500011875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181852.hg.1 |
TC0500011875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011875.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
ENST00000306442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
ENST00000415659.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
NM_000943.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181853.hg.1 |
TC0500011875.hg.1 |
Clariom D TC |