Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003934.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003934.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003934.hg.1 |
TC17000323.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000577936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007372.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
ENST00000226247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
ENST00000579795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
NM_003593.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152432.hg.1 |
TC1700007372.hg.1 |
Clariom D TC |