Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010215.hg.1 |
ENST00000284384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010215.hg.1 |
ENST00000413366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010215.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010215.hg.1 |
TC17000783.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164755.hg.1 |
TC1700008661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000284384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000413366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000578063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
ENST00000583775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008661.hg.1 |
NM_002737.2 |
Refseq |