Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012147.hg.1 |
ENST00000393094.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012147.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012147.hg.1 |
TC11000977.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000531843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166261.hg.1 |
TC1100008979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000393094.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000526303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000531843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
ENST00000532782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008979.hg.1 |
NM_003478.4 |
Refseq |