Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030096.hg.1 |
TC07001937.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030096.hg.1 |
ENST00000547058.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030096.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030096.hg.1 |
NM_001001317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030098.hg.1 |
TC07001937.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030098.hg.1 |
ENST00000547058.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030098.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030098.hg.1 |
NM_001001317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700192702.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192702.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192702.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192702.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192702.hg.1 |
TC0700012855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700192705.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192705.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192705.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192705.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192705.hg.1 |
TC0700012855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700192703.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192703.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192703.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192703.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192703.hg.1 |
TC0700012855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000547058.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012855.hg.1 |
ENST00000552471.1 |
Ensembl |