Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006083.hg.1 |
TC0X000471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006083.hg.1 |
ENST00000450049.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006083.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000372953.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000450457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
ENST00000494863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NM_001162491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007930.hg.1 |
NR_103709.1 |
Refseq |