Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008341.hg.1 |
TC04000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008341.hg.1 |
ENST00000265162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008341.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008340.hg.1 |
TC04000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008340.hg.1 |
ENST00000265162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008340.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008342.hg.1 |
TC04000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008342.hg.1 |
ENST00000265162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008342.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008343.hg.1 |
TC04000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008343.hg.1 |
ENST00000265162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008343.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000502711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000504100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012836.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188787.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188787.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188787.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188787.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188787.hg.1 |
TC0400012836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188789.hg.1 |
TC0400012836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
ENST00000509344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188788.hg.1 |
TC0400012836.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400188786.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188786.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188786.hg.1 |
ENST00000265162.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188786.hg.1 |
NM_001977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188786.hg.1 |
TC0400012836.hg.1 |
Clariom D TC |