Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207419.3 | NP_997302.2 | 3 - 4 | 483 | 90 | |
XM_011522483.2 | XP_011520785.1 | 3 - 4 | 566 | 90 | ||
GenBank mRNA | AY358832.1 | - | 3 - 4 | 483 | 90 | |
FJ422561.1 | - | 2 - 3 | 253 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |