Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
ENST00000002501.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
ENST00000304733.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
ENST00000392973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018859.hg.1 |
TC16001361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194913.hg.1 |
TC1600011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194908.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194908.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194908.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194908.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194908.hg.1 |
TC1600011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194912.hg.1 |
TC1600011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194901.hg.1 |
TC1600011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000002501.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000304733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000392973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000568838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
ENST00000623401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001042610.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288708.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_001288709.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011585.hg.1 |
NM_024043.3 |
Refseq |