Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011988.hg.1 |
ENST00000539521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008363.hg.1 |
TC0X000636.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008363.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008363.hg.1 |
ENST00000370796.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008363.hg.1 |
ENST00000462974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008363.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008359.hg.1 |
TC0X000636.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008359.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008359.hg.1 |
ENST00000370796.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008359.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008360.hg.1 |
TC0X000636.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008360.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008360.hg.1 |
ENST00000370796.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008360.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177939.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177939.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177939.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177939.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177939.hg.1 |
TC0X00011326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177940.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177940.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177940.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177940.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177940.hg.1 |
TC0X00011326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177941.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177941.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177941.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177941.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177941.hg.1 |
TC0X00011326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011988.hg.1 |
ENST00000539521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177944.hg.1 |
TC0X00011326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000298556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
ENST00000462974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011326.hg.1 |
NM_000194.2 |
Refseq |