Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000465325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013809.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000396203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000416518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000435633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000461368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000465280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000479563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
ENST00000481798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
NM_001034996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
NM_003973.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203726.hg.1 |
TC0300013809.hg.1 |
Clariom D TC |