Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006442.hg.1 |
TC05000456.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006442.hg.1 |
ENST00000283357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006442.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006438.hg.1 |
TC05000456.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006438.hg.1 |
ENST00000283357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006438.hg.1 |
ENST00000506418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006438.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006443.hg.1 |
TC05000456.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006443.hg.1 |
ENST00000283357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006443.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000512365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008114.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155521.hg.1 |
TC0500008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155522.hg.1 |
TC0500008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000283357.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000503099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000503361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000506418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000507832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000510458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
ENST00000513529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
NM_152548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155517.hg.1 |
TC0500008114.hg.1 |
Clariom D TC |