Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004437.hg.1 |
TC09000415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004437.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004437.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153023.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153023.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153023.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153023.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153023.hg.1 |
TC0900007863.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153021.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153021.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153021.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153021.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153021.hg.1 |
TC0900007863.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |