Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
ENST00000255688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
ENST00000439013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006937.hg.1 |
TC11000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156332.hg.1 |
TC1100007899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000255688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000354445.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000439013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000537871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
ENST00000544107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007899.hg.1 |
NM_004585.4 |
Refseq |