Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178056.1 | NP_001171527.1 | 15 - 16 | 1817 | 117 | |
XM_011543632.2 | XP_011541934.2 | 16 - 17 | 1777 | 117 | ||
GenBank mRNA | BC037386.1 | - | 13 - 14 | 1675 | 4830820 | 117 |
BC075801.1 | - | 15 - 16 | 1776 | 5226828 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |