Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020777.hg.1 |
TC06001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020777.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020777.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020778.hg.1 |
TC06001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020778.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020778.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020776.hg.1 |
TC06001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020776.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020776.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181346.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181346.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181346.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181346.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181346.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000244670.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000326974.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
ENST00000332245.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NM_057161.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008066.hg.1 |
NR_040101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181347.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181347.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181347.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181347.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181347.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600181349.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181349.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181349.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181349.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181349.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600181350.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181350.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181350.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181350.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181350.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011853.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181345.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181345.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181345.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181345.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181345.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600181348.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181348.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181348.hg.1 |
ENST00000244711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181348.hg.1 |
NM_014623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181348.hg.1 |
TC0600011853.hg.1 |
Clariom D TC |