Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001874.hg.1 |
ENST00000377191.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001874.hg.1 |
ENST00000430571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001874.hg.1 |
ENST00000539513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001874.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001874.hg.1 |
TC20000140.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000147246.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147246.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147246.hg.1 |
TC2000006899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147246.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147246.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
ENST00000377191.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006899.hg.1 |
NM_012255.4 |
Refseq |