Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028732.hg.1 |
TC01001848.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028732.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028732.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240465.hg.1 |
TC0100018349.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000272133.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000470788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000478120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000479227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000481095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000489556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000492470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000496277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
ENST00000498382.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018349.hg.1 |
NM_152495.1 |
Refseq |