Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000294053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000340729.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000437826.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000535477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000535990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000538021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000538039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000543042.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258393.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_030813.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000294053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000340729.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000437826.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000535477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000535990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000538021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000538039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
ENST00000543042.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258393.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_001258394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
NM_030813.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188962.hg.1 |
TC1100011552.hg.1 |
Clariom D TC |