Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
TC01001185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
ENST00000414970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
ENST00000467329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
ENST00000543398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018199.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000467514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000470824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000476970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000493553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
ENST00000496202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009902.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
ENST00000324862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
ENST00000467329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
NM_004698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171501.hg.1 |
TC0100009902.hg.1 |
Clariom D TC |