Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016352.hg.1 |
TC04001194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016352.hg.1 |
ENST00000381431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016352.hg.1 |
ENST00000535450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016352.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016349.hg.1 |
TC04001194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016349.hg.1 |
ENST00000381431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016349.hg.1 |
ENST00000535450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016349.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016351.hg.1 |
TC04001194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016351.hg.1 |
ENST00000381431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016351.hg.1 |
ENST00000535450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016351.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016350.hg.1 |
TC04001194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016350.hg.1 |
ENST00000381431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016350.hg.1 |
ENST00000535450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016350.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010682.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172737.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172737.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172737.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172737.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172737.hg.1 |
TC0400010682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172736.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172736.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172736.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172736.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172736.hg.1 |
TC0400010682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172738.hg.1 |
TC0400010682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000381431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000506357.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
ENST00000514133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
NM_000232.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172739.hg.1 |
TC0400010682.hg.1 |
Clariom D TC |