Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123.3 | NP_001114.2 | 2 - 3 | 479 | 132 | |
NM_001202450.1 | NP_001189379.1 | 2 - 3 | 240 | 132 | ||
NM_006721.3 | NP_006712.2 | 2 - 3 | 240 | 132 | ||
XM_017015698.1 | XP_016871187.1 | 2 - 3 | 240 | 132 | ||
XM_017015700.1 | XP_016871189.1 | 2 - 3 | 240 | 132 | ||
XM_017015701.1 | XP_016871190.1 | 2 - 3 | 414 | 132 | ||
XM_017015702.1 | XP_016871191.1 | 2 - 3 | 414 | 132 | ||
XM_017015704.1 | XP_016871193.1 | 2 - 3 | 240 | 132 | ||
XM_017015705.1 | XP_016871194.1 | 2 - 3 | 240 | 132 | ||
XM_017015706.1 | XP_016871195.1 | 2 - 3 | 414 | 132 | ||
GenBank mRNA | AK290633.1 | - | 2 - 3 | 205 | 132 | |
AK301590.1 | - | 2 - 3 | 223 | 132 | ||
BC003568.1 | - | 2 - 3 | 194 | 3544131 | 132 | |
BM472149.1 | - | 2 - 3 | 479 | 132 | ||
BT006788.1 | - | 1 - 2 | 95 | 132 | ||
CB133126.1 | - | 2 - 3 | 218 | 132 | ||
DA356691.1 | - | 2 - 3 | 241 | 132 | ||
U33936.1 | - | 2 - 3 | 153 | 132 | ||
U50196.1 | - | 2 - 3 | 278 | 132 | ||
U90338.1 | - | 2 - 3 | 196 | 132 | ||
U90339.1 | - | 2 - 3 | 188 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |