Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000580712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002683.hg.1 |
TC04000175.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002683.hg.1 |
ENST00000302874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002683.hg.1 |
ENST00000450254.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002683.hg.1 |
ENST00000505451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002683.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002692.hg.1 |
TC04000175.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002692.hg.1 |
ENST00000302874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002692.hg.1 |
ENST00000450254.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002692.hg.1 |
ENST00000505451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002692.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148572.hg.1 |
TC0400007095.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148573.hg.1 |
TC0400007095.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000302874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000505451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000507760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000508058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000580712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
ENST00000612982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007095.hg.1 |
NM_024936.2 |
Refseq |