Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018384.hg.1 |
ENST00000268616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018384.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018384.hg.1 |
TC16001332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018385.hg.1 |
ENST00000268616.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018385.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018385.hg.1 |
TC16001332.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186805.hg.1 |
TC1600011168.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000565193.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011168.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000268616.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000561928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
ENST00000568020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
NM_015144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186803.hg.1 |
TC1600011168.hg.1 |
Clariom D TC |