Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002000.hg.1 |
TC07000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002000.hg.1 |
ENST00000222584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002000.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000440636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006864.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000222584.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000432066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
ENST00000448246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
NM_003112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147437.hg.1 |
TC0700006864.hg.1 |
Clariom D TC |