Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000442.hg.1 |
ENST00000321153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000442.hg.1 |
ENST00000530797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000442.hg.1 |
ENST00000532004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000442.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000442.hg.1 |
TC11000030.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000362915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006491.hg.1 |
NR_002585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000441.hg.1 |
ENST00000321153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000441.hg.1 |
ENST00000530797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000441.hg.1 |
ENST00000532004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000441.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000441.hg.1 |
TC11000030.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144896.hg.1 |
TC1100006491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144901.hg.1 |
TC1100006491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000362915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000525722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000526222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000527517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
ENST00000532004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006491.hg.1 |
NR_002585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000321153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000524867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
ENST00000530797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
NM_001004.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144893.hg.1 |
TC1100006491.hg.1 |
Clariom D TC |