Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004685.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004685.hg.1 |
ENST00000375547.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004685.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004685.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004685.hg.1 |
TC13000418.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154601.hg.1 |
TC1300008111.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000342783.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000375547.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
ENST00000493630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_001256134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008111.hg.1 |
NM_003891.2 |
Refseq |