Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015384.4 | NP_056199.2 | 16 - 17 | 4343 | 115 | |
NM_133433.3 | NP_597677.2 | 16 - 17 | 4343 | 115 | ||
XM_005248280.3 | XP_005248337.1 | 16 - 17 | 4332 | 115 | ||
XM_005248282.4 | XP_005248339.3 | 10 - 11 | 3834 | 115 | ||
XM_006714467.2 | XP_006714530.1 | 16 - 17 | 4332 | 115 | ||
XM_006714468.2 | XP_006714531.1 | 15 - 16 | 4134 | 115 | ||
XM_011514015.1 | XP_011512317.1 | 16 - 17 | 4332 | 115 | ||
XM_017009329.1 | XP_016864818.1 | 16 - 17 | 4332 | 115 | ||
XM_017009330.1 | XP_016864819.1 | 8 - 9 | 6606 | 115 | ||
XM_017009331.1 | XP_016864820.1 | 15 - 16 | 2706 | 115 | ||
GenBank mRNA | AJ627032.1 | - | 16 - 17 | 4312 | 115 | |
AJ640137.1 | - | 16 - 17 | 4312 | 115 | ||
KJ807791.1 | - | 2 - 3 | 130 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |