Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009399.hg.1 |
ENST00000376250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009399.hg.1 |
ENST00000455100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009399.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009399.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009399.hg.1 |
TC13000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
ENST00000376250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
ENST00000455100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
ENST00000492399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009401.hg.1 |
TC13000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009396.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009396.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009396.hg.1 |
TC13000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009398.hg.1 |
ENST00000376250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009398.hg.1 |
ENST00000455100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009398.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009398.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009398.hg.1 |
TC13000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164387.hg.1 |
TC1300009637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000454752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000464500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000467518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000471912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000492399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000608779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
ENST00000629479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009637.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
ENST00000376250.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
ENST00000455100.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
NM_001195087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
NM_033110.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164386.hg.1 |
TC1300009637.hg.1 |
Clariom D TC |